siRNA Details
| Gene Symbol: Rac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134366.1 | NP_599193.1 | 2, 3 | 110 | ||
| XM_008768986.1 | XP_008767208.1 | 2, 3 | 144 | |||
| GenBank mRNA | AF385833.1 | - | 1, 2 | 38 | ||
| AY491395.1 | - | 2, 3 | 110 | |||

Rac1
Genomic Map