siRNA Details
| Gene Symbol: Tmem132c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002727968.4 | XP_002728014.2 | 2 | 1093 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem132c| Gene Symbol: Tmem132c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002727968.4 | XP_002728014.2 | 2 | 1093 | ||