siRNA Details
| Gene Symbol: Rad9b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001030042.1 | NP_001025213.1 | 9 | 952 | ||
| XM_006249366.2 | XP_006249428.1 | 9 | 1578 | |||
| XM_008769242.1 | XP_008767464.1 | 9 | 970 | |||
| XM_008769243.1 | XP_008767465.1 | 8 | 771 | |||
| XM_008769244.1 | XP_008767466.1 | 9 | 866 | |||
| XM_008769245.1 | XP_008767467.1 | 9 | 805 | |||
| XM_008769246.1 | XP_008767468.1 | 9 | 799 | |||
| GenBank mRNA | BC099750.1 | - | 9 | 952 | 5599441 | |

Rad9b
Genomic Map