siRNA Details
| Gene Symbol: Rad9b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001030042.1 | NP_001025213.1 | 3 | 286 | ||
| XM_006249366.2 | XP_006249428.1 | 3 | 912 | |||
| XM_008769241.1 | XP_008767463.1 | 3 | 300 | |||
| XM_008769242.1 | XP_008767464.1 | 3 | 304 | |||
| XM_008769243.1 | XP_008767465.1 | 2 | 105 | |||
| XM_008769244.1 | XP_008767466.1 | 3 | 200 | |||
| XM_008769245.1 | XP_008767467.1 | 3 | 139 | |||
| XM_008769246.1 | XP_008767468.1 | 3 | 133 | |||
| GenBank mRNA | BC099750.1 | - | 3 | 286 | 5599441 | |

Rad9b
Genomic Map