siRNA Details
| Gene Symbol: Ncf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100984.1 | NP_001094454.1 | 9 | 809 | ||
| XM_006250013.2 | XP_006250075.1 | 8 | 1028 | |||
| XM_008769664.1 | XP_008767886.1 | 6 | 800 | |||
| GenBank mRNA | JN864041.1 | - | 7 | 833 | ||
| JN864042.1 | - | 7 | 833 | |||
| JN864045.1 | - | 7 | 827 | |||
| JN864046.1 | - | 6 | 719 | |||

Ncf2
Genomic Map