siRNA Details
| Gene Symbol: Mael | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108857.1 | NP_001102327.1 | 7, 8 | 776 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mael| Gene Symbol: Mael | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108857.1 | NP_001102327.1 | 7, 8 | 776 | ||