siRNA Details
| Gene Symbol: Kctd8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100172.1 | NP_001093642.1 | 1 | 1051 | ||
| GenBank mRNA | EF043041.1 | - | 1 | 1051 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kctd8| Gene Symbol: Kctd8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100172.1 | NP_001093642.1 | 1 | 1051 | ||
| GenBank mRNA | EF043041.1 | - | 1 | 1051 | ||