siRNA Details
| Gene Symbol: Dnajc9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108865.1 | NP_001102335.1 | 3 | 464 | ||
| GenBank mRNA | AB002591.1 | - | 3 | 715 | ||
| FQ215794.1 | - | 3 | 458 | |||
| FQ229094.1 | - | 3 | 446 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dnajc9| Gene Symbol: Dnajc9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108865.1 | NP_001102335.1 | 3 | 464 | ||
| GenBank mRNA | AB002591.1 | - | 3 | 715 | ||
| FQ215794.1 | - | 3 | 458 | |||
| FQ229094.1 | - | 3 | 446 | |||