siRNA Details
| Gene Symbol: Abhd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108866.1 | NP_001102336.1 | 2, 3 | 180 | ||
| XM_006251995.2 | XP_006252057.1 | 2, 3 | 385 | |||
| XM_006251996.2 | XP_006252058.1 | 2, 3 | 176 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Abhd4| Gene Symbol: Abhd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108866.1 | NP_001102336.1 | 2, 3 | 180 | ||
| XM_006251995.2 | XP_006252057.1 | 2, 3 | 385 | |||
| XM_006251996.2 | XP_006252058.1 | 2, 3 | 176 | |||