siRNA Details
| Gene Symbol: Prmt5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108867.2 | NP_001102337.2 | 12, 13 | 1379 | ||
| GenBank mRNA | CB793692.1 | - | 1, 2 | 169 | ||
| DY553859.1 | - | 6, 7 | 633 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prmt5| Gene Symbol: Prmt5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108867.2 | NP_001102337.2 | 12, 13 | 1379 | ||
| GenBank mRNA | CB793692.1 | - | 1, 2 | 169 | ||
| DY553859.1 | - | 6, 7 | 633 | |||