siRNA Details
| Gene Symbol: Prmt5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108867.2 | NP_001102337.2 | 5, 6 | 575 | ||
| GenBank mRNA | CO573950.1 | - | 3, 4 | 375 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prmt5| Gene Symbol: Prmt5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108867.2 | NP_001102337.2 | 5, 6 | 575 | ||
| GenBank mRNA | CO573950.1 | - | 3, 4 | 375 | ||