siRNA Details
| Gene Symbol: Prmt5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108867.2 | NP_001102337.2 | 14, 15 | 1590 | ||
| GenBank mRNA | BQ210423.1 | - | 3, 4 | 668 | ||
| CB793692.1 | - | 3, 4 | 380 | |||
| CK845516.1 | - | 3, 4 | 668 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prmt5| Gene Symbol: Prmt5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108867.2 | NP_001102337.2 | 14, 15 | 1590 | ||
| GenBank mRNA | BQ210423.1 | - | 3, 4 | 668 | ||
| CB793692.1 | - | 3, 4 | 380 | |||
| CK845516.1 | - | 3, 4 | 668 | |||