siRNA Details
| Gene Symbol: Kctd9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108871.1 | NP_001102341.1 | 1, 2 | 128 | ||
| XM_006252138.2 | XP_006252200.1 | 1, 2 | 114 | |||
| XM_006252139.2 | XP_006252201.1 | 1, 2 | 233 | |||
| GenBank mRNA | FQ221845.1 | - | 1, 2 | 238 | ||

Kctd9
Genomic Map