siRNA Details
| Gene Symbol: Tlr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198791.1 | NP_942086.1 | 6 | 2579 | ||
| XM_008771266.1 | XP_008769488.1 | 7 | 2828 | |||
| XM_008771267.1 | XP_008769489.1 | 7 | 2748 | |||
| GenBank mRNA | AB116229.1 | - | 6 | 2579 | ||

Tlr3
Genomic Map