siRNA Details
| Gene Symbol: Tlr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198791.1 | NP_942086.1 | 4 | 385 | ||
| XM_008771266.1 | XP_008769488.1 | 5 | 634 | |||
| XM_008771267.1 | XP_008769489.1 | 5 | 554 | |||
| GenBank mRNA | AB116229.1 | - | 4 | 385 | ||

Tlr3
Genomic Map