siRNA Details
| Gene Symbol: Fbxo25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014239.1 | NP_001014261.1 | 7 | 770 | ||
| GenBank mRNA | BC082088.1 | - | 7 | 770 | 7133991 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fbxo25| Gene Symbol: Fbxo25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014239.1 | NP_001014261.1 | 7 | 770 | ||
| GenBank mRNA | BC082088.1 | - | 7 | 770 | 7133991 | |