siRNA Details
| Gene Symbol: B4galt7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031661.1 | NP_001026831.1 | 2 | 139 | ||
| XM_008771500.1 | XP_008769722.1 | 2 | 1896 | |||
| XM_008771501.1 | XP_008769723.1 | 3 | 339 | |||
| GenBank mRNA | BC099103.1 | - | 2 | 139 | 7460148 | |

B4galt7
Genomic Map