siRNA Details
| Gene Symbol: Olr1657 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000536.1 | NP_001000536.1 | 1 | 88 | ||
| Gene Symbol: LOC102557552 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008771774.1 | XP_008769996.1 | 1 | 141 | ||
| Gene Symbol: LOC103690297 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008771775.1 | XP_008769997.1 | 1 | 105 | ||

Olr1657
Genomic Map