siRNA Details
| Gene Symbol: Nme8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198792.2 | NP_942087.2 | 16 | 1848 | ||
| XM_006254088.2 | XP_006254150.1 | 16 | 1897 | |||
| GenBank mRNA | AF548544.1 | - | 14 | 1555 | ||
| BC097265.1 | - | 15 | 1622 | |||

Nme8
Genomic Map