siRNA Details
| Gene Symbol: Itga8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001173972.1 | NP_001167443.1 | 18 | 1973 | ||
| GenBank mRNA | BC168655.1 | - | 18 | 1973 | 7122577 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Itga8| Gene Symbol: Itga8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001173972.1 | NP_001167443.1 | 18 | 1973 | ||
| GenBank mRNA | BC168655.1 | - | 18 | 1973 | 7122577 | |