siRNA Details
| Gene Symbol: Rnaseh2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013234.1 | NP_001013252.1 | 4, 5 | 894 | ||
| XM_008772414.1 | XP_008770636.1 | 4, 5 | 424 | |||
| XM_008772415.1 | XP_008770637.1 | 2, 3 | 208 | |||
| XM_008772416.1 | XP_008770638.1 | 2, 3 | 204 | |||
| GenBank mRNA | BC086345.1 | - | 4, 5 | 424 | 7302028 | |
| BC086539.1 | - | 4, 5 | 894 | 7114943 | ||
| FQ227287.1 | - | 2, 3 | 253 | |||

Rnaseh2a
Genomic Map