siRNA Details
| Gene Symbol: Zfp324 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008758845.1 | XP_008757067.1 | 2 | 1167 | ||
| XM_008758846.1 | XP_008757068.1 | 2 | 1141 | |||
| XM_008758847.1 | XP_008757069.1 | 2 | 1494 | |||
| XM_344862.7 | XP_344863.6 | 4 | 883 | |||

Zfp324
Genomic Map