siRNA Details
| Gene Symbol: RGD1561430 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006228847.2 | XP_006228909.1 | 1 | 722 | ||
| XM_008759258.1 | XP_008757480.1 | 1 | 722 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1561430| Gene Symbol: RGD1561430 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006228847.2 | XP_006228909.1 | 1 | 722 | ||
| XM_008759258.1 | XP_008757480.1 | 1 | 722 | |||