siRNA Details
| Gene Symbol: Alpk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191895.1 | NP_001178824.1 | 3, 4 | 294 | ||
| XM_008759546.1 | XP_008757768.1 | 2, 3 | 301 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Alpk3| Gene Symbol: Alpk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191895.1 | NP_001178824.1 | 3, 4 | 294 | ||
| XM_008759546.1 | XP_008757768.1 | 2, 3 | 301 | |||