siRNA Details
| Gene Symbol: Jakmip3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163277.1 | NP_001156749.1 | 5 | 1184 | ||
| XM_008760004.1 | XP_008758226.1 | 9 | 2279 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Jakmip3| Gene Symbol: Jakmip3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163277.1 | NP_001156749.1 | 5 | 1184 | ||
| XM_008760004.1 | XP_008758226.1 | 9 | 2279 | |||