siRNA Details
| Gene Symbol: Lrrc56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024902.1 | NP_001020073.1 | 8 | 810 | ||
| XM_006230564.2 | XP_006230626.1 | 8 | 1296 | |||
| XM_006230565.2 | XP_006230627.1 | 8 | 1406 | |||
| XM_006230566.2 | XP_006230628.1 | 8 | 1579 | |||
| XM_006230569.2 | XP_006230631.1 | 8 | 1333 | |||
| XM_006230570.2 | XP_006230632.1 | 6 | 543 | |||
| XM_008760005.1 | XP_008758227.1 | 8 | 1578 | |||
| GenBank mRNA | BC097444.1 | - | 8 | 810 | 7456428 | |

Lrrc56
Genomic Map