siRNA Details
| Gene Symbol: Osbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108927.1 | NP_001102397.1 | 3, 4 | 720 | ||
| XM_006231090.1 | XP_006231152.1 | 3, 4 | 1040 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Osbp| Gene Symbol: Osbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108927.1 | NP_001102397.1 | 3, 4 | 720 | ||
| XM_006231090.1 | XP_006231152.1 | 3, 4 | 1040 | |||