siRNA Details
Gene Symbol: Osbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108927.1 | NP_001102397.1 | 3 | 557 | ||
XM_006231090.1 | XP_006231152.1 | 3 | 877 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Osbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108927.1 | NP_001102397.1 | 3 | 557 | ||
XM_006231090.1 | XP_006231152.1 | 3 | 877 |