siRNA Details
| Gene Symbol: Cfap43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006231588.2 | XP_006231650.1 | 10, 11 | 1206 | ||
| XM_345041.7 | XP_345042.5 | 14, 15 | 1994 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cfap43| Gene Symbol: Cfap43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006231588.2 | XP_006231650.1 | 10, 11 | 1206 | ||
| XM_345041.7 | XP_345042.5 | 14, 15 | 1994 | |||