siRNA Details
| Gene Symbol: Cfap43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006231588.2 | XP_006231650.1 | 2 | 190 | ||
| XM_008760497.1 | XP_008758719.1 | 6 | 978 | |||
| XM_345041.7 | XP_345042.5 | 6 | 978 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cfap43| Gene Symbol: Cfap43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006231588.2 | XP_006231650.1 | 2 | 190 | ||
| XM_008760497.1 | XP_008758719.1 | 6 | 978 | |||
| XM_345041.7 | XP_345042.5 | 6 | 978 | |||