siRNA Details
| Gene Symbol: Dse | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108933.1 | NP_001102403.1 | 6 | 2110 | ||
| XM_006256382.2 | XP_006256444.1 | 6 | 1865 | |||
| XM_008772900.1 | XP_008771122.1 | 6 | 1942 | |||
| XM_008772901.1 | XP_008771123.1 | 8 | 2444 | |||
| GenBank mRNA | BC168891.1 | - | 8 | 2378 | 7110121 | |

Dse
Genomic Map