siRNA Details
| Gene Symbol: Zfr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270972.1 | NP_001257901.1 | 5 | 972 | ||
| XM_006232054.2 | XP_006232116.1 | 4 | 766 | |||
| XM_008760782.1 | XP_008759004.1 | 4 | 1028 | |||
| XM_008760783.1 | XP_008759005.1 | 5 | 967 | |||

Zfr
Genomic Map