siRNA Details
| Gene Symbol: Hnf4g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108939.1 | NP_001102409.1 | 8 | 1166 | ||
| XM_006232165.2 | XP_006232227.1 | 8 | 1917 | |||
| XM_006232166.2 | XP_006232228.1 | 9 | 1760 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hnf4g| Gene Symbol: Hnf4g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108939.1 | NP_001102409.1 | 8 | 1166 | ||
| XM_006232165.2 | XP_006232227.1 | 8 | 1917 | |||
| XM_006232166.2 | XP_006232228.1 | 9 | 1760 | |||