siRNA Details
| Gene Symbol: Fstl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271195.1 | NP_001258124.1 | 7 | 1575 | ||
| XM_008761106.1 | XP_008759328.1 | 4 | 477 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fstl5| Gene Symbol: Fstl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271195.1 | NP_001258124.1 | 7 | 1575 | ||
| XM_008761106.1 | XP_008759328.1 | 4 | 477 | |||