siRNA Details
| Gene Symbol: Nup210l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191900.1 | NP_001178829.1 | 38 | 5422 | ||
| XM_006232724.2 | XP_006232786.1 | 38 | 5479 | |||
| XM_006232726.2 | XP_006232788.1 | 35 | 5086 | |||
| XM_008761195.1 | XP_008759417.1 | 37 | 5347 | |||
| XM_008761196.1 | XP_008759418.1 | 36 | 5029 | |||
| XM_008761197.1 | XP_008759419.1 | 34 | 5023 | |||
| XM_008761198.1 | XP_008759420.1 | 37 | 5277 | |||
| XM_008761199.1 | XP_008759421.1 | 36 | 5029 | |||

Nup210l
Genomic Map