siRNA Details
| Gene Symbol: Ankrd35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006232996.2 | XP_006233058.1 | 9 | 1411 | ||
| XM_345258.7 | XP_345259.5 | 11 | 1681 | |||
| GenBank mRNA | FQ221348.1 | - | 10 | 1585 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ankrd35| Gene Symbol: Ankrd35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006232996.2 | XP_006233058.1 | 9 | 1411 | ||
| XM_345258.7 | XP_345259.5 | 11 | 1681 | |||
| GenBank mRNA | FQ221348.1 | - | 10 | 1585 | ||