siRNA Details
| Gene Symbol: Trim33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006233100.2 | XP_006233162.1 | 18 | 3099 | ||
| XM_345266.7 | XP_345267.3 | 18 | 3099 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trim33| Gene Symbol: Trim33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006233100.2 | XP_006233162.1 | 18 | 3099 | ||
| XM_345266.7 | XP_345267.3 | 18 | 3099 | |||