siRNA Details
| Gene Symbol: Bank1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047918.2 | NP_001041383.2 | 12 | 2050 | ||
| GenBank mRNA | FQ231577.1 | - | 11 | 1730 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bank1| Gene Symbol: Bank1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047918.2 | NP_001041383.2 | 12 | 2050 | ||
| GenBank mRNA | FQ231577.1 | - | 11 | 1730 | ||