siRNA Details
| Gene Symbol: Nutm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108957.1 | NP_001102427.1 | 2 | 510 | ||
| XM_006234708.2 | XP_006234770.1 | 2 | 506 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nutm1| Gene Symbol: Nutm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108957.1 | NP_001102427.1 | 2 | 510 | ||
| XM_006234708.2 | XP_006234770.1 | 2 | 506 | |||