siRNA Details
| Gene Symbol: Rtf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108958.1 | NP_001102428.1 | 17 | 1953 | ||
| XM_006234783.1 | XP_006234845.1 | 18 | 2100 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rtf1| Gene Symbol: Rtf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108958.1 | NP_001102428.1 | 17 | 1953 | ||
| XM_006234783.1 | XP_006234845.1 | 18 | 2100 | |||