siRNA Details
| Gene Symbol: Catsper2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012220.1 | NP_001012220.1 | 5 | 789 | ||
| XM_008762187.1 | XP_008760409.1 | 5 | 471 | |||
| GenBank mRNA | BC079422.1 | - | 5 | 789 | 7114001 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Catsper2| Gene Symbol: Catsper2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012220.1 | NP_001012220.1 | 5 | 789 | ||
| XM_008762187.1 | XP_008760409.1 | 5 | 471 | |||
| GenBank mRNA | BC079422.1 | - | 5 | 789 | 7114001 | |