siRNA Details
Gene Symbol: Ntsr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108967.1 | NP_001102437.1 | 1, 2 | 1171 | ||
XM_006235788.1 | XP_006235850.1 | 1, 2 | 1171 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
