siRNA Details
| Gene Symbol: Prpf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079766.1 | NP_001073234.1 | 3 | 368 | ||
| GenBank mRNA | BC128779.1 | - | 3 | 368 | 7191091 | |
| FQ221925.1 | - | 3 | 371 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prpf6| Gene Symbol: Prpf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079766.1 | NP_001073234.1 | 3 | 368 | ||
| GenBank mRNA | BC128779.1 | - | 3 | 368 | 7191091 | |
| FQ221925.1 | - | 3 | 371 | |||