siRNA Details
| Gene Symbol: RGD1310641 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079701.1 | NP_001073169.1 | 12, 13 | 1768 | ||
| GenBank mRNA | BC128759.1 | - | 12, 13 | 1768 | 7112639 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1310641| Gene Symbol: RGD1310641 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079701.1 | NP_001073169.1 | 12, 13 | 1768 | ||
| GenBank mRNA | BC128759.1 | - | 12, 13 | 1768 | 7112639 | |