siRNA Details
| Gene Symbol: Slc5a9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108974.1 | NP_001102444.1 | 4 | 489 | ||
| XM_006238598.2 | XP_006238660.1 | 6 | 764 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc5a9| Gene Symbol: Slc5a9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108974.1 | NP_001102444.1 | 4 | 489 | ||
| XM_006238598.2 | XP_006238660.1 | 6 | 764 | |||