siRNA Details
| Gene Symbol: Spata21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004447.1 | NP_001004447.1 | 12 | 1838 | ||
| XM_008764277.1 | XP_008762499.1 | 12 | 1987 | |||
| GenBank mRNA | AB168097.1 | - | 12 | 1838 | ||
| BC089089.1 | - | 11 | 1757 | 7112067 | ||

Spata21
Genomic Map