siRNA Details
| Gene Symbol: Tnfrsf14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001015034.1 | NP_001015034.1 | 6 | 647 | ||
| XM_006239586.2 | XP_006239648.1 | 7 | 1532 | |||
| XM_006239587.2 | XP_006239649.1 | 7 | 1533 | |||
| GenBank mRNA | BC091202.1 | - | 6 | 647 | 7380378 | |

Tnfrsf14
Genomic Map