siRNA Details
| Gene Symbol: Atp6v1e2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108979.1 | NP_001102449.1 | 2 | 619 | ||
| XM_006239715.2 | XP_006239777.1 | 2 | 665 | |||
| XM_006239716.2 | XP_006239778.1 | 2 | 1066 | |||
| XM_008764465.1 | XP_008762687.1 | 2 | 642 | |||
| XM_008764466.1 | XP_008762688.1 | 2 | 681 | |||

Atp6v1e2
Genomic Map