siRNA Details
Gene Symbol: Slc30a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013243.1 | NP_001013261.1 | 6 | 929 | ||
XM_008764525.1 | XP_008762747.1 | 7 | 929 | |||
XM_008764526.1 | XP_008762748.1 | 7 | 974 | |||
XM_008764527.1 | XP_008762749.1 | 7 | 903 | |||
XM_008764528.1 | XP_008762750.1 | 7 | 1009 | |||
XM_008764529.1 | XP_008762751.1 | 8 | 1104 | |||
GenBank mRNA | AY538655.1 | - | 6 | 929 | ||
BC086513.1 | - | 7 | 901 | 7134069 |
