siRNA Details
| Gene Symbol: Slc30a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013243.1 | NP_001013261.1 | 8 | 1155 | ||
| XM_008764525.1 | XP_008762747.1 | 9 | 1155 | |||
| XM_008764526.1 | XP_008762748.1 | 9 | 1200 | |||
| XM_008764527.1 | XP_008762749.1 | 9 | 1129 | |||
| XM_008764528.1 | XP_008762750.1 | 9 | 1235 | |||
| XM_008764529.1 | XP_008762751.1 | 10 | 1330 | |||
| GenBank mRNA | AY538655.1 | - | 8 | 1155 | ||
| BC086513.1 | - | 9 | 1127 | 7134069 | ||

Slc30a3
Genomic Map